Learn More
Aviva Systems Biology Corp LRFN3 Antibody - C-terminal region (ARP49822_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49822_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LRFN3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human LRFN3. Alternate names: FIGLER1, MGC2656, SALM4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60195