Learn More
Aviva Systems Biology Corp LRFN5 Antibody - middle region (ARP52979_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52979_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LRFN5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LRFN5. Alternate names: C14orf146, DKFZp686G0210, FIGLER8, FLJ30803, SALM5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53701