Learn More
Essendant Mop Head, Looped, Enviro Clean With Tailband, Large, White, 12/Carton
Catalog No. 502004313
Encompass_Preferred
Patented, mildew-resistant DuPont material is designed to meet clean room (Class 1000) applications for lint reduction. Ideal for cleanup of foodservice areas, hospitals and oil and hazardous chemical spills. 100% recycled, cotton/synthetic blend fibers are interlocked for strength, launderability and less linting. Fibertac-processed cut-end mops last three times longer than comparable cut-end mops. Durable, double-sewn tailband.
Catalog No. 50-200-4313 Supplier Essendant Supplier No. BWK8003
Patented, mildew-resistant DuPont material is designed to meet clean room (Class 1000) applications for lint reduction. Ideal for cleanup of foodservice areas, hospitals and oil and hazardous chemical spills. 100% recycled, cotton/synthetic blend fibers are interlocked for strength, launderability and less linting. Fibertac-processed cut-end mops last three times longer than comparable cut-end mops. Durable, double-sewn tailband.