Learn More
Aviva Systems Biology Corp LRG1 Antibody - N-terminal region (ARP52475_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52475_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LRG1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human LRG1. Alternate names: HMFT1766, LRGApplicationWB. Species reactivity: Cow, Human, Pig
Catalog No. 50-285-719