Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC LRGILS-NH2 TFA | 98.9% | 770.84 | 10 MG
SDP

Catalog No. 5000423508
Encompass_Preferred

LRGILS-NH2 TFA is a reverse-sequence protease-activated receptor-2 (PAR-2)-inactive, negative control peptide, in contrast to SLIGRL-NH2, which is a PAR-2-activating peptide. It is intended for research use only.

  • Reverse-sequence protease-activated receptor-2 (PAR-2)-inactive
  • Functions as a negative control peptide
  • Solid appearance, white to off-white in color
  • Sequence: Leu-Arg-Gly-Ile-Leu-Ser-NH2
  • Soluble in DMSO (100 mg/mL)

Catalog No. 50-004-23508 Supplier Medchemexpress LLC Supplier No. HYP1312A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LRGILS-NH2 TFA is a reverse-sequence protease-activated receptor-2 (PAR-2)-inactive, negative control peptide, in contrast to SLIGRL-NH2, which is a PAR-2-activating peptide. It is intended for research use only.

  • Reverse-sequence protease-activated receptor-2 (PAR-2)-inactive
  • Functions as a negative control peptide
  • Solid appearance, white to off-white in color
  • Sequence: Leu-Arg-Gly-Ile-Leu-Ser-NH2
  • Soluble in DMSO (100 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.