Learn More
Medchemexpress LLC (+)-LRH-1 modulator-1 | 00-00-0 | 99.4% | C28H36N2O2S | 10MG
SDP
Catalog No. 5000258646
Encompass_Preferred
(+)-LRH-1 modulator-1 is the (+)-stereoisomer of an LRH-1 modulator and functions as a potent agonist of liver receptor homolog-1 (LRH-1). It modulates inflammatory signaling in intestinal organoid models by inducing IL-10 and reducing IL-1β and TNFα. The compound is provided as a high-purity research-grade small molecule for cellular and biochemical studies of LRH-1 pathways.
  • Potent LRH-1 modulator/agonist for receptor pharmacology studies.
  • Induces anti-inflammatory cytokine IL-10 in organoid models.
  • Reduces proinflammatory cytokines IL-1β and TNFα in assays.
  • High purity suitable for cellular and biochemical experiments.
  • Available in multiple small-scale quantities for research use.
Catalog No. 50-002-58646 Supplier Medchemexpress LLC Supplier No. HY147105A10MG
(+)-LRH-1 modulator-1 is the (+)-stereoisomer of an LRH-1 modulator and functions as a potent agonist of liver receptor homolog-1 (LRH-1). It modulates inflammatory signaling in intestinal organoid models by inducing IL-10 and reducing IL-1β and TNFα. The compound is provided as a high-purity research-grade small molecule for cellular and biochemical studies of LRH-1 pathways.
  • Potent LRH-1 modulator/agonist for receptor pharmacology studies.
  • Induces anti-inflammatory cytokine IL-10 in organoid models.
  • Reduces proinflammatory cytokines IL-1β and TNFα in assays.
  • High purity suitable for cellular and biochemical experiments.
  • Available in multiple small-scale quantities for research use.