Learn More
Medchemexpress LLC LRH-1 modulator-1 | 2244781-29-9 | 98.0% | C28H36N2O2S | 10MG
SDP
Catalog No. 5000261604
Encompass_Preferred
LRH-1 modulator-1 is a research-grade small molecule characterized as a potent agonist/modulator of the liver receptor homolog-1 (LRH-1). The product is provided with analytical documentation and recommended storage and solubility guidance for laboratory and preclinical use.
  • Potent LRH-1 modulator/agonist for receptor biology studies.
  • High purity (98.0%) suitable for analytical and biological assays.
  • Supplied with certificate of analysis, HNMR, and LCMS data for verification.
  • Soluble in DMSO; in vivo formulation protocols are provided.
  • Stable under specified storage conditions with solvent-specific guidelines.
Catalog No. 50-002-61604 Supplier Medchemexpress LLC Supplier No. HY14710510MG
LRH-1 modulator-1 is a research-grade small molecule characterized as a potent agonist/modulator of the liver receptor homolog-1 (LRH-1). The product is provided with analytical documentation and recommended storage and solubility guidance for laboratory and preclinical use.
  • Potent LRH-1 modulator/agonist for receptor biology studies.
  • High purity (98.0%) suitable for analytical and biological assays.
  • Supplied with certificate of analysis, HNMR, and LCMS data for verification.
  • Soluble in DMSO; in vivo formulation protocols are provided.
  • Stable under specified storage conditions with solvent-specific guidelines.