Learn More
Medchemexpress LLC (+)-LRH-1 modulator-1 | 2244781-29-9 | 99.38% | 464.66 | 50 MG
SDP
Catalog No. 5000362186
Encompass_Preferred
(+)-LRH-1 modulator-1 is a stereoisomer of LRH-1 modulator-1, a potent LRH-1 (liver receptor homolog-1) modulator/agonist. It exhibits anti-inflammatory effects in intestinal organoids by inducing the anti-inflammatory cytokine IL-10 and reducing the inflammatory cytokines IL-1b and TNFa.
  • Potent LRH-1 modulator/agonist
  • Exhibits anti-inflammatory effects in intestinal organoids
  • Induces anti-inflammatory cytokine IL-10
  • Reduces inflammatory cytokines IL-1b and TNFa
  • Pure form storage: -20°C for 3 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Shipping conditions: Room temperature if less than 2 weeks
Catalog No. 50-003-62186 Supplier Medchemexpress LLC Supplier No. HY147105A50MG
(+)-LRH-1 modulator-1 is a stereoisomer of LRH-1 modulator-1, a potent LRH-1 (liver receptor homolog-1) modulator/agonist. It exhibits anti-inflammatory effects in intestinal organoids by inducing the anti-inflammatory cytokine IL-10 and reducing the inflammatory cytokines IL-1b and TNFa.
  • Potent LRH-1 modulator/agonist
  • Exhibits anti-inflammatory effects in intestinal organoids
  • Induces anti-inflammatory cytokine IL-10
  • Reduces inflammatory cytokines IL-1b and TNFa
  • Pure form storage: -20°C for 3 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Shipping conditions: Room temperature if less than 2 weeks