Learn More
Medchemexpress LLC LRH-1 modulator-1 | 2244781-29-9 | 99.4% | 464.66 | 5 MG
SDP
Catalog No. 5000362377
Encompass_Preferred
(+)-LRH-1 modulator-1 is a stereoisomer of LRH-1 modulator-1. LRH-1 modulator-1 (compound 6N) is a potent LRH-1 (liver receptor homolog-1) modulator/agonist. It has anti-inflammatory effects in intestinal organoids, inducing the anti-inflammatory cytokine IL-10 and reducing the inflammatory cytokines IL-1b and TNFa.
  • Potent LRH-1 modulator/agonist
  • Anti-inflammatory effects in intestinal organoids
  • Induces anti-inflammatory cytokine IL-10
  • Reduces inflammatory cytokines IL-1b and TNFa
Catalog No. 50-003-62377 Supplier Medchemexpress LLC Supplier No. HY147105A5MG
(+)-LRH-1 modulator-1 is a stereoisomer of LRH-1 modulator-1. LRH-1 modulator-1 (compound 6N) is a potent LRH-1 (liver receptor homolog-1) modulator/agonist. It has anti-inflammatory effects in intestinal organoids, inducing the anti-inflammatory cytokine IL-10 and reducing the inflammatory cytokines IL-1b and TNFa.
  • Potent LRH-1 modulator/agonist
  • Anti-inflammatory effects in intestinal organoids
  • Induces anti-inflammatory cytokine IL-10
  • Reduces inflammatory cytokines IL-1b and TNFa