Learn More
ACROBiosystems ACROBiosystems Human LRP-5 Protein, Mouse IgG2a Fc Tag
Catalog No. 501956821
Encompass_Preferred
Recombinant Protein;1MG;Human LRP-5, Mouse IgG2a Fc Tag (LR5-H5254) is expressed from human 293 cells (HEK293). It contains AA Glu 644 - Gln 1263 (Accession # O75197-1 ).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 96.6 kDa. The protein migrates as 110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human Dkk-1, Fc,Avitag (Cat. No. DK1-H82F5) at 5 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Human LRP-5, Mouse IgG2a Fc Tag (Cat. No. LR5-H5254) with a linear range of 0.5-16 ng/mL (QC tested).;Immobilized Human Dkk-1, His Tag (Cat. No. DK1-H5221) at 5 μg/mL (100 μL/well) can bind Human LRP-5, Mouse IgG2a Fc Tag (Cat. No. LR5-H5254) with a linear range of 0.01-0.313 μg/mL (Routinely tested).
Catalog No. 50-195-6821 Supplier ACROBiosystems Supplier No. LR5H52541MG
Recombinant Protein;1MG;Human LRP-5, Mouse IgG2a Fc Tag (LR5-H5254) is expressed from human 293 cells (HEK293). It contains AA Glu 644 - Gln 1263 (Accession # O75197-1 ).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 96.6 kDa. The protein migrates as 110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human Dkk-1, Fc,Avitag (Cat. No. DK1-H82F5) at 5 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Human LRP-5, Mouse IgG2a Fc Tag (Cat. No. LR5-H5254) with a linear range of 0.5-16 ng/mL (QC tested).;Immobilized Human Dkk-1, His Tag (Cat. No. DK1-H5221) at 5 μg/mL (100 μL/well) can bind Human LRP-5, Mouse IgG2a Fc Tag (Cat. No. LR5-H5254) with a linear range of 0.01-0.313 μg/mL (Routinely tested).