Learn More
ACROBiosystems ACROBiosystems Human LRP-6 (20-630) Protein, Mouse IgG2a Fc Tag
Supplier:  ACROBiosystems LR6H5253100UG
Encompass_Preferred
Recombinant Protein;100UG;Human LRP-6 (20-630), Mouse IgG2a Fc Tag (LR6-H5253) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Pro 630 (Accession # O75581-1 ).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 95.8 kDa. The protein migrates as 55 kDa and 110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse Dkk-1, His Tag (Cat. No. DK1-M52H6) at 5 μg/mL (100 μL/well) can bind Human LRP-6 (20-630), Mouse IgG2a Fc Tag (Cat. No. LR6-H5253) with a linear range of 10-78 ng/mL (QC tested).
Catalog No. 50-195-6709