Learn More
Aviva Systems Biology Corp LRP1 Antibody - middle region (ARP58563_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58563_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LRP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LRP1. Alternate names: APR, LRP, A2MR, CD91, APOER, TGFBR5, IGFBP3RApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62356