Learn More
Aviva Systems Biology Corp Lrp2bp Antibody - middle region (ARP42516_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42516_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Lrp2bp. It was validated on Western Blot. . Alternate names: RGD1305896ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-008