Learn More
Aviva Systems Biology Corp LRP2BP Antibody - middle region (ARP42515_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42515_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LRP2BP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LRP2BP. Alternate names: DKFZp761O0113ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53343