Learn More
Aviva Systems Biology Corp Lrp8 Antibody - middle region (ARP44306_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44306_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Lrp8. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 4932703M08Rik, AA921429, AI848122, Lr8b, apoER2, ApoER2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-285-135