Learn More
Aviva Systems Biology Corp LRP8 Antibody - middle region (ARP44723_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44723_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LRP8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LRP8. Alternate names: APOER2, HSZ75190, MCI1, LRP-8ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-58885