Learn More
Aviva Systems Biology Corp LRPAP1 Antibody - C-terminal region (ARP45276_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45276_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against LRPAP1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human LRPAP1. Alternate names: A2MRAP, A2RAP, HBP44, MGC138272, MRAP, RAPApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-58996