Learn More
Cell Signaling Technology LRRC15 Rabbit mAb, Specific For Total LRRC15 Protein, Validated For WB, IHC-P, 100 µl
SDP
Supplier:  Cell Signaling Technology 50546S
Encompass
The LRRC15 (E4X8J) Rabbit Monoclonal Antibody is a validated tool for detecting endogenous total LRRC15 protein, crucial for cancer research. It offers high specificity and reliability in Western Blot and Immunohistochemistry applications. With its consistent performance across lots, this antibody serves as a dependable choice, particularly for studies on TGF-β-driven cancer-associated fibroblasts, representing a potential therapeutic target in various solid tumors.
Catalog No. NC2818609