Learn More
Aviva Systems Biology Corp LRRC20 Antibody - middle region (ARP57172_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57172_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LRRC20. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LRRC20. Alternate names: FLJ10751, FLJ10844ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62098