Learn More
Aviva Systems Biology Corp LRRC23 Antibody - middle region (ARP53487_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53487_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LRRC23. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human LRRC23. Alternate names: LRPB7ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-842