Learn More
Aviva Systems Biology Corp LRRC24 Antibody - N-terminal region (ARP44738_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44738_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LRRC24. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LRRC24. Alternate names: LRRC14OS, MGC111484ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-58888