Learn More
Aviva Systems Biology Corp LRRC26 Antibody - middle region (ARP44660_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44660_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LRRC26. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LRRC26. Alternate names: bA350O14.10, CAPCApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human
Catalog No. 50-014-53430