Learn More
Aviva Systems Biology Corp LRRC3 Antibody - middle region (ARP45878_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45878_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LRRC3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human LRRC3. Alternate names: C21orf102ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-223