Learn More
Medchemexpress LLC LRRC32 beta 50ug | 50ug
SDP
Catalog No. 5000187075
Encompass_Preferred
LRRC32 is a key regulator of TGF- activation and maintains TGFB1 TGFB2 and TGFB3 in the latent state during extracellular storage by binding to latency-associated peptide (LAP) LRRC32 competes with LTBP1 for LAP binding and effectively regulates integrin-dependent TGF- activation GARP Latent TGF beta Complex Protein Human (Biotinylated HEK293 His-Avi) is a recombinant protein dimer complex containing human-derived GARP Latent TGF beta Complex protein expressed by HEK293 with C-Avi C-His labeled tag GARP Latent TGF beta Complex Protein Human (Biotinylated HEK293 His-Avi) has molecular weight of (73-78) kDa (GARP) 13 kDa (42-45) kDa (Latent TGF beta 1) respectively
Catalog No. 50-001-87075 Supplier Medchemexpress LLC Supplier No. HYP7767550UG
LRRC32 is a key regulator of TGF- activation and maintains TGFB1 TGFB2 and TGFB3 in the latent state during extracellular storage by binding to latency-associated peptide (LAP) LRRC32 competes with LTBP1 for LAP binding and effectively regulates integrin-dependent TGF- activation GARP Latent TGF beta Complex Protein Human (Biotinylated HEK293 His-Avi) is a recombinant protein dimer complex containing human-derived GARP Latent TGF beta Complex protein expressed by HEK293 with C-Avi C-His labeled tag GARP Latent TGF beta Complex Protein Human (Biotinylated HEK293 His-Avi) has molecular weight of (73-78) kDa (GARP) 13 kDa (42-45) kDa (Latent TGF beta 1) respectively