Learn More
ACROBiosystems ACROBiosystems Human LRRC32 / GARP / Garpin Protein, Fc Tag
Catalog No. 501957163
Encompass_Preferred
Recombinant Protein;1MG;Human LRRC32, Fc Tag (LR2-H5256) is expressed from human 293 cells (HEK293). It contains AA His 20 - Asn 627 (Accession # Q14392-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 92.4 kDa. The protein migrates as 105 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7163 Supplier ACROBiosystems Supplier No. LR2H52561MG
Recombinant Protein;1MG;Human LRRC32, Fc Tag (LR2-H5256) is expressed from human 293 cells (HEK293). It contains AA His 20 - Asn 627 (Accession # Q14392-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 92.4 kDa. The protein migrates as 105 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.