Learn More
Enzo Life Sciences LRRC32 (human) : Fc (human), (recombinant) (50µg)
SDP
Supplier:  Enzo Life Sciences ALX522117C050
Encompass_Preferred
Alternative name: Leucine rich repeat containing protein 32, GARP, Garpin, Glycoprotein A repetitions predominant. Concentration: 1mg/ml (after reconstitution). Purity: ≥90% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in CHO cells. Human LRRC32 (aa 20-627) is fused to the Fc portion of human IgG1. Long Term Storage: -20°C.
Catalog No. 50-200-9437