Learn More
Enzo Life Sciences LRRC32 monoclonal antibody (Plato-1) (FITC conjugate) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804867FI0100
Encompass_Preferred
Clone: Plato-1. Isotype: IgG2b. Immunogen: Recombinant human LRRC32 (aa 20-627). Source: Purified from concentrated hybridoma tissue culture supernatant. Species reactivity: Human, Mouse. Applications: Flow Cytometry. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 0.09% sodium azide.
Catalog No. 50-201-0009