Learn More
ACROBiosystems Recombinant Protein;Human LRRC32 / GARP / Garpin Protein, Fc Tag;HEK293;100UG;LR2,LRRC32,GARP,Garpin,D11S833E
Supplier: ACROBiosystems LR2H5256100UG
Recombinant Protein;Human LRRC32 / GARP / Garpin Protein, Fc Tag;100UG;Human LRRC32, Fc Tag (LR2-H5256) is expressed from human 293 cells (HEK293). It contains AA His 20 - Asn 627 (Accession # Q14392-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 92.4 kDa. The protein migrates as 106 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TGFB1, His Tag (Cat. No. TG1-H524x) at 5 μg/mL (100 μL/well) can bind Human LRRC32, Fc Tag (Cat. No. LR2-H5256) with a linear range of 0.078-1.25 μg/mL (QC tested).