Learn More
Aviva Systems Biology Corp LRRC33 Antibody - N-terminal region (ARP55907_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55907_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LRRC33. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LRRC33. Alternate names: GARPL1, MGC50789, UNQ3030, LRRC33ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61659