Learn More
Aviva Systems Biology Corp LRRC37A3 Antibody - middle region (ARP56070_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56070_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LRRC37A3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LRRC37A3. Alternate names: FLJ34306, KIAA0563, MGC41826, MGC57168, LRRC37AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-61705