Learn More
Sino Biological LRRC3B Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 15120T32100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human LRRC3B (rh LRRC3B; Catalog#15120-H08H; NP_443185.1; Met1-Tyr204). LRRC3B specific IgG was purified by Human LRRC3B affinity chromatography.
Catalog No. 50-224-8060