Learn More
Aviva Systems Biology Corp LRRC4C Antibody - N-terminal region (ARP49593_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49593_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LRRC4C. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LRRC4C. Alternate names: KIAA1580, NGL-1, NGL1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60120