Learn More
ACROBiosystems Recombinant Protein;Human LRRC4 Protein;HEK293;1MG;LR4,LRRC4,NGL-2,BAG,NAG14,NGL2,LRRC-4,Netrin-G2 ligand
Catalog No. 501658009
Encompass_Preferred
Recombinant Protein;Human LRRC4 Protein;1MG;Human LRRC4, His Tag (LR4-H5221) is expressed from human 293 cells (HEK293). It contains AA Ala 39 - Lys 527 (Accession # AAI11562.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 55.6 kDa. The protein migrates as 90-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8009 Supplier ACROBiosystems Supplier No. LR4H52211MG
Recombinant Protein;Human LRRC4 Protein;1MG;Human LRRC4, His Tag (LR4-H5221) is expressed from human 293 cells (HEK293). It contains AA Ala 39 - Lys 527 (Accession # AAI11562.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 55.6 kDa. The protein migrates as 90-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.