Learn More
Aviva Systems Biology Corp LRRC56 Antibody - N-terminal region (ARP53452_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53452_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LRRC56. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LRRC56. Alternate names: DKFZp761L1518, FLJ00101ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-61029