Learn More
Aviva Systems Biology Corp LRRC57 Antibody - middle region (ARP55578_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55578_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LRRC57. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LRRC57. Alternate names: DKFZp686H1865, FLJ36812ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61544