Learn More
Aviva Systems Biology Corp LRRC66 Antibody - middle region (ARP44734_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44734_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC339977. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LRRC66. Alternate names: ApplicationWB. Species reactivity: Human
Catalog No. 50-014-53907