Learn More
Aviva Systems Biology Corp LRRC7 Antibody - C-terminal region (ARP64881_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64881_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LRRC7. It was validated on Western Blot. . Alternate names: DENSINApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-385