Learn More
Aviva Systems Biology Corp LRRC8A Antibody - N-terminal region (ARP49452_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49452_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LRRC8A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LRRC8A. Alternate names: FLJ10337, KIAA1437, LRRC8, AGM5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53584