Learn More
Aviva Systems Biology Corp LRRC8B Antibody - middle region (ARP47145_P050) | 100uL
SDP
Catalog No. 5001453506
Encompass_Preferred
This is a rabbit polyclonal antibody against LRRC8B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LRRC8B. Alternate names: KIAA0231, MGC42220, TA-LRRP, TALRRPApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53506 Supplier Aviva Systems Biology Corp Supplier No. ARP47145_P050
This is a rabbit polyclonal antibody against LRRC8B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LRRC8B. Alternate names: KIAA0231, MGC42220, TA-LRRP, TALRRPApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat