Learn More
Aviva Systems Biology Corp LRRFIP1 Antibody - middle region (ARP59016_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59016_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LRRFIP1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of Human LRRFIP1. Alternate names: FLAP-1, FLAP1, FLIIAP1, GCF-2, GCF2, HUFI-1, TRIPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit
Catalog No. 50-286-559