Learn More
Aviva Systems Biology Corp LRRFIP2 Antibody - N-terminal region (ARP59017_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59017_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LRRFIP2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human LRRFIP2. Alternate names: DKFZp434H2035, FLJ20248, FLJ22683, FLJ58304, HUFI-2ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-286-560