Learn More
Eurofins Discoverx LRRK2, active, 10 µg. (14-919)
Supplier:  Eurofins Discoverx 14919
Encompass
N-terminal FLAG-tagged recombinant, human LRRK2 amino acids 970-end. This protein is a member of the leucine-rich repeat kinase family and encodes a protein with an ankryin repeat region, a leucine-rich repeat (LRR) domain, a kinase domain, a DFG-like motif, a RAS domain, a GTPase domain, a MLK-like domain, and a WD40 domain. The protein is present largely in the cytoplasm but also associates with the mitochondrial outer membrane. Mutations in this gene have been associated with Parkinson disease-8. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9413