Learn More
Abcam LRRK2-IN-1, LRRK2 inhibitor, 50MG
Catalog No. 502672387
Encompass_Preferred
MW 570.7 Da. Potent and selective inhibitor of the Parkinson's disease kinase LRRK2. Benzodiazepine based derivative. Inhibits both G2019S mutant and wild-type LRRK2 kinase activity (IC₅₀ values are 6 and 13 nM respectively). Causes dephosphorylation, ubiquitination and degradation of LRRK2. Inhibits IFN-γ-induced monocyte maturation in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-2387 Supplier Abcam Supplier No. AB25452450MG
MW 570.7 Da. Potent and selective inhibitor of the Parkinson's disease kinase LRRK2. Benzodiazepine based derivative. Inhibits both G2019S mutant and wild-type LRRK2 kinase activity (IC₅₀ values are 6 and 13 nM respectively). Causes dephosphorylation, ubiquitination and degradation of LRRK2. Inhibits IFN-γ-induced monocyte maturation in vitro.
The product is subject to the following: Abcam Restricted Use Statement