Learn More
Sino Biological Recombinant Human c-Kit Protein (d557-558, GST Tag), Active
Supplier:  Sino Biological K0612WG50
Encompass_Preferred
Recombinant human c-KIT (d557-558) (544-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-4551