Learn More
Enzo Life Sciences LRRKtide (1mg)
SDP
Catalog No. 502011324
Encompass_Preferred
This peptide, RLGRDKYKTLRQIRQ, is derived from ezrin/moesin/radixin and is a substrate for LRRK kinase. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-1324 Supplier Enzo Life Sciences Supplier No. BMLP2670001
This peptide, RLGRDKYKTLRQIRQ, is derived from ezrin/moesin/radixin and is a substrate for LRRK kinase. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water. Long Term Storage: -20°C.