Learn More
Enzo Life Sciences LRRKtide (biotinylated) (1mg)
SDP
Catalog No. 502011325
Encompass_Preferred
This peptide, Biotin-Ahx-RLGRDKYKTLRQIRQ, is derived from ezrin/moesin/radixin and is a substrate for LRRK kinase. The biotin allows the peptide to be used in kinase assays with streptavidin-bound membranes. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-1325 Supplier Enzo Life Sciences Supplier No. BMLP2680001
This peptide, Biotin-Ahx-RLGRDKYKTLRQIRQ, is derived from ezrin/moesin/radixin and is a substrate for LRRK kinase. The biotin allows the peptide to be used in kinase assays with streptavidin-bound membranes. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water. Long Term Storage: -20°C.