Learn More
Cayman Chemical LRRKtIDtrIfluoROCTAte sa 10mg
SDP
Catalog No. 502917835
Encompass_Preferred
A peptide substrate for LRRK2; has been used to identify inhibitors of LRRK2
Catalog No. 50-291-7835 Supplier Cayman Chemical Supplier No. 3746110 MG
A peptide substrate for LRRK2; has been used to identify inhibitors of LRRK2