Learn More
Aviva Systems Biology Corp LRRTM4 Antibody - middle region (ARP49915_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49915_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LRRTM4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LRRTM4. Alternate names: FLJ12568, MGC120633, MGC120636ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60213