Learn More
STEMCELL Technologies LRS Cone, Size: 1 Unit
SDP
Catalog No. NC1861457
Encompass
Leukocyte Reduction System (LRS) cones are used during the collection of leukapheresis products to reduce the leukocyte count from human blood collections. These products collect the leukocytes, which can then be further processed using cell separation products, such as EasySep™ Cell Isolation Kits, to purify cell populations for study. Cells were obtained using Institutional Review Board (IRB)-approved consent forms and protocols.
Catalog No. NC1861457 Supplier STEMCELL Technologies Supplier No. 2000093
Leukocyte Reduction System (LRS) cones are used during the collection of leukapheresis products to reduce the leukocyte count from human blood collections. These products collect the leukocytes, which can then be further processed using cell separation products, such as EasySep™ Cell Isolation Kits, to purify cell populations for study. Cells were obtained using Institutional Review Board (IRB)-approved consent forms and protocols.