Learn More
Teledyne ISCO, Inc Ls Class Single Piston Pump With Belt Drive And Shaped Cam; 0.0 - 40.0 Ml/Min. ; 1,600 Psi
Catalog No. 502119949 Shop All Teledyne ISCO Inc. Products
Encompass
LS Class Single Piston Pump with Belt Drive and Shaped Cam; 0.0 - 40.0 mL/min. ; 1,600 psi; PEEK Fluid Path; Automatic Piston Wash; with Pulse Dampener; Standard Seal (Flouropolymer energizer); Fitting package for 1/8"""" and 1/16"""" tubing
Catalog No. 50-211-9949 Supplier Teledyne ISCO, Inc Supplier No. LS040PRX
Add to Cart
Add to Cart
Request a Quote
LS Class Single Piston Pump with Belt Drive and Shaped Cam; 0.0 - 40.0 mL/min. ; 1,600 psi; PEEK Fluid Path; Automatic Piston Wash; with Pulse Dampener; Standard Seal (Flouropolymer energizer); Fitting package for 1/8"""" and 1/16"""" tubing